Få mere at vide
Urea-d4, TRC
Artikelnummer. 30455906
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30455906 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455906 Leverandør Toronto Research Chemicals Leverandørnr. TRCU8225061G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Urea-d4
CAS 1433-11-0
Molekylær formel C D4 N2 O
Mængde 1 g
Synonym Urea-d4 (6CI,7CI,8CI,9CI), 1,1,3,3-Tetradeuteriourea, Urea-d4, Urea-D4, Carbamimidic acid-d4, Carbamimidic-d4 acid, Carbonyl diamide-d4, Urea D4, [2H4]-Urea
InChI formel InChI=1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i/hD4
SMIL [2H]N([2H])C(=O)N([2H])[2H]
IUPAC navn 1,1,3,3-tetradeuteriourea
Molekylvægt (g/mol) 64.08
Formel vægt 64.057