Få mere at vide
Urea-d4, CDN
Artikelnummer. 30319206
Skift visning
Mængde:
25 g
Artikelnummer. Mængde
30319206 25 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319206 Leverandør CDN Leverandørnr. CDND039125G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Urea-d4
CAS 1433-11-0
Molekylær formel C D4 N2 O
Mængde 25 g
Synonym Urea-d4 (6 CI,7 CI,8 CI,9 CI), 1,1,3,3-Tetradeuteriourea, Urea-d4, Urea-D4, Carbamimidic acid-d4, Carbamimidic-d4 acid, Carbonyl diamide-d4, Urea D4, [2H4]-Urea
InChI formel InChI=1 S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i/hD4
SMIL [2 H]N([2 H])C(=O)N([2 H])[2 H]
IUPAC navn 1,1,3,3-tetradeuteriourea
Molekylvægt (g/mol) 64.08
Formel vægt 64.057 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity