Få mere at vide
URB602, MedChemExpress
Artikelnummer. 16499716
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16499716 50 mg
16483168 10 mg
16444456 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16499716 Leverandør Medchem Express Leverandørnr. HY100792
MedChemExpress URB602 is a selective monoacylglycerol lipase (MGL) inhibitor, which inhibits rat brain MGL with IC50 of 28±4 μM through a noncompetitive mechanism.
URB602 is a selective monoacylglycerol lipase (MGL) inhibitor, which inhibits rat brain MGL with IC50 of 28±4 μM through a noncompetitive mechanism.
Tekniske data
Kemisk navn eller materiale URB602
Fysisk form Solid
CAS 565460-15-3
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C19H21NO2
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : 100 mg/mL (338.55 mM; Need ultrasonic)
SMIL O=C(OC1CCCCC1)NC2=CC(C3=CC=CC=C3)=CC=C2
Molekylvægt (g/mol) 295.38
Formel vægt 295.38
Procent renhed 99.51%