Få mere at vide
Urapidil-d3 (methoxy-d3), CDN
Artikelnummer. 30324360
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324360 0.01 g
30322918 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324360 Leverandør CDN Leverandørnr. CDND73730.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Urapidil-D3 (methoxy-D3)
CAS 1398066-08-4
Molekylær formel C20 D3 H26 N5 O3
Mængde 0.01 g
Synonym Urapidil-D3 (methoxy-D3), Urapidil-d3, 6-[[3-[4-[2-(Methoxy-d3)phenyl]-1-piperazinyl]propyl]amino]-1,3-dimethyl-2,4(1 H,3 H)-pyrimidinedione
InChI formel InChI=1 S/C20H29N5O3/c1-22-18(15-19(26)23(2)20(22)27)21-9-6-10-24-11-13-25(14-12-24)16-7-4-5-8-17(16)28-3/h4-5,7-8,15,21 H,6,9-14H2,1-3H3/i3D3
SMIL [2 H]C([2 H])([2 H])Oc1ccccc1N2CCN(CCCNC3=CC(=O)N(C)C(=O)N3C)CC2
IUPAC navn 1,3-dimethyl-6-[3-[4-[2-(trideuteriomethoxy)phenyl]piperazin-1-yl]propylamino]pyrimidine-2,4-dione
Molekylvægt (g/mol) 390.494
Formel vægt 390.246 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity