Få mere at vide
Uracil-13C,15N2, TRC
Artikelnummer. 30004124
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004124 50 mg
30011776 25 mg
30011730 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004124 Leverandør Toronto Research Chemicals Leverandørnr. TRCU80100250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181517-11-3
Molekylær formel C313CH415N2O2
Molekylvægt (g/mol) 115.07
Synonym 2,4(1H,3H)-Pyrimidinedione-13C,15N2, 2,4-Dihydroxypyrimidine-13C,15N2, 2,4-Dioxopyrimidine-13C,15N2, 2,4-Pyrimidinediol-13C,15N2, 2,4-Pyrimidinedione-13C,15N2, 4-Hydroxyuracil-13C,15N2, Hybar X-13C,15N2;
IUPAC navn pyrimidine-2,4(1H,3H)-dione-2-13C-1,3-15N2
SMIL O=[13C]([15NH]1)[15NH]C=CC1=O
Tekniske data
Kemisk navn eller materiale Uracil-13C,15N2
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)/i4+1,5+1,6+1
Formel vægt 115.02
Procent renhed >95%