Få mere at vide
Uracil-1,3-d2, CDN
Artikelnummer. 30310143
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310143 0.25 g -
30317118 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310143 Leverandør CDN Leverandørnr. CDND18830.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Uracil-1,3-D2
CAS 20666-60-8
Molekylær formel C4 D2 H2 N2 O2
Mængde 0.25 g
Synonym 2,4(1 H,3 H)-Pyrimidinedione-1,3-d2 (9 CI, ACI), Uracil-1,3-d2 (7 CI, 8 CI), 1,3-Dideuteriouracil, 1,3-Dideuterouracil, [1,3-d2]Uracil, Uracil-1,3-D2
InChI formel InChI=1 S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2 H,(H2,5,6,7,8)/i/hD2
SMIL [2 H]N1C=CC(=O)N([2 H])C1=O
IUPAC navn 1,3-dideuteriopyrimidine-2,4-dione
Molekylvægt (g/mol) 114.099
Formel vægt 114.04 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity