Learn More
Kinesis™ Inlet Solvent Filters: End Fittings
Manufactured from Polyether ether ketone (PEEK). Upchurch Scientific Inlet Solvent Filters: End Fittings are used for Analytical HPLC Systems.
1050.00 DKK
Specifications
Type | Replacement Filter End Fitting |
---|---|
Material | PEEK |
Thread Size | 10 to 32 Coned |
Description
- For Analytical HPLC System
- 0.5μm PEEK filter end fittings
- Black body
- Swept volume of 5.7μL
- Thread port: 10-32 Coned
- For Analytical HPLC System
- 0.5μm or 0.2μm PEEK filter end fittings
- Black or natural body
- Swept volume of 5.7μL or 6.9μL
- Thread port: 10-32 Coned
Specifications
Replacement Filter End Fitting | |
PEEK | |
10 to 32 Coned |