Få mere at vide
Upadacitinib, MedChemExpress
Artikelnummer. 16493656
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
500 mg
Pakningsstørrelse:
10mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16493656 25 mg -
16473767 50 mg -
16450278 5 mg 5mg
16447646 100 mg -
16417818 10 mg 10mg
16411906 500 mg -
16411296 200 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Artikelnummer. 16493656 Leverandør Medchem Express Leverandørnr. HY19569100mg
MedChemExpress Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relevant cytokines. Upadacitinib (ABT-494) can be used for several autoimmune disorders research.
Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relevant cytokines. Upadacitinib (ABT-494) can be used for several autoimmune disorders research.
Tekniske data
Kemisk navn eller materiale Upadacitinib
Fysisk form Powder
CAS 1310726-60-3
Farve Gray
Mængde 25 mg
Renhedsgrad noter Research
Molekylær formel C17H19F3N6O
Til brug med (applikation) COVID-19-immunoregulation
Synonym ABT-494
Opløselighedsinformation DMSO : ≥ 22 mg/mL (57.84 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(N1C[C@@H](CC)[C@@H](C2=CN=C3C=NC(NC=C4)=C4N32)C1)NCC(F)(F)F
Molekylvægt (g/mol) 380.37
Formel vægt 380.37
Procent renhed 98.56%