Learn More
Alexandra™ Unisex White Stud Front Lab Coat
Product Code. 15851144
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15851144 Small Each
15801144 2X-Large Each
15811144 3X-Large Each
15821144 4X-Large Each
15831144 X-Small Each
15851134 Medium Each
15861134 Large Each
15881134 X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15851144 Supplier Alexandra™ Supplier No. G178WHS
  • ONLINE EXCLUSIVE PRICE
    290.14 DKK
    197.82 DKK / Pakke med 1
    Spare 92.32 DKK
    31% Slukket
Forventet forsendelse: 22-12-2026
Machine-washable, polyester/cotton, unisex asymmetrical stud-front laboratory coats feature stretch cuffs for comfort and three useful pockets.
  • Unisex asymmetrical stud front laboratory coat
  • Stretch cuffs for comfort
  • Two hip pockets, one chest pocket
  • Machine washable and industrially launderable
  • Unisex
  • Length 43ins/110cm
  • 245gsm 67% polyester/33% cotton
  • Available in a choice of colours in the wider product portfolio
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polycotton
Color White
Closure Type Press Studs
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Mandarin
Size (English) Chest 34 in.
Includes Assymetric stud front, two hip and one chest pockets
Length (English) 43 inch
Length (Metric) 110 cm