Learn More
Alexandra™ Unisex White Stud Front Lab Coat
Product Code. 15831144
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15831144 X-Small Each
15801144 2X-Large Each
15811144 3X-Large Each
15821144 4X-Large Each
15851134 Medium Each
15851144 Small Each
15861134 Large Each
15881134 X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15831144 Supplier Alexandra™ Supplier No. G178WHXS
  • ONLINE EXCLUSIVE PRICE
    290.14 DKK
    177.89 DKK / Pakke med 1
    Spare 112.25 DKK
    38% Slukket
Forventet forsendelse: 08-01-2027
Machine-washable, polyester/cotton, unisex asymmetrical stud-front laboratory coats feature stretch cuffs for comfort and three useful pockets.
  • Unisex asymmetrical stud front laboratory coat
  • Stretch cuffs for comfort
  • Two hip pockets, one chest pocket
  • Machine washable and industrially launderable
  • Unisex
  • Length 43ins/110cm
  • 245gsm 67% polyester/33% cotton
  • Available in a choice of colours in the wider product portfolio
Specifications
Product Type General Purpose Lab Coat
Size X-Small
Gender Unisex
Material Polycotton
Color White
Closure Type Press Studs
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Mandarin
Size (English) Chest 32 in.
Includes Assymetric stud front, two hip and one chest pockets
Length (English) 43 inch
Length (Metric) 110 cm