Learn More
Masterflex™ UNF Male Threaded Adapters, PVDF
Combine these simple, economical PVDF male threaded adaptors with flexible tubing for use in your low-pressure applications (generally less than 30 psi). UNF Male Threaded Adapters slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
Brand:  Masterflex™ 30704-88
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13224450
  • 211.00 DKK / Pack of 10
Estimated Shipment
24-05-2021
Add to basket
Description
Description
Combine these simple, economical PVDF male threaded adaptors with flexible tubing for use in your low-pressure applications (generally less than 30 psi). UNF Male Threaded Adapters slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
  • Adapts from a UNF male thread to a hose barb
  • Designed for quick and easy assembly without tools
  • Excellent chemical resistance
  • Temp range: -62 to 129°C
  • Sterilize by ethylene oxide or autoclave
Specifications
Specifications
Autoclavable
Barbed Fitting
10 Pack
1/4 to 28 UNF(M)
PVDF
1/4 to 28 UNF(M) thread x 3/16 in. tubing I.D.
1/8 in. orifice; 15/16 x 3/8 in.