Få mere at vide
Undecanoic-d21 Acid, TRC
Artikelnummer. 30449059
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30449059 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449059 Leverandør Toronto Research Chemicals Leverandørnr. TRCU78885150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Undecanoic-D21 Acid
CAS 60658-40-4
Molekylær formel C11HD21O2
Mængde 50 mg
Synonym Undecanoic-d21 acid (9CI), Undecanoic acid D21, Undecanoic acid-d21, Perdeuteroundecanoic acid
InChI formel InChI=1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O
IUPAC navn undecanoic-d21 acid
Molekylvægt (g/mol) 207.42
Formel vægt 207.29