Få mere at vide
Undecanoic-11,11,11-d3 Acid, CDN
Artikelnummer. 30319960
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319960 0.1 g
30322724 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319960 Leverandør CDN Leverandørnr. CDND60020.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Undecanoic-11,11,11-D3 Acid
CAS 1219802-11-5
Molekylær formel C11 D3 H19 O2
Mængde 0.1 g
Synonym 11,11,11-Trideuteroundecanoic acid, ω-Trideuteroundecanoic acid, Undecanoic acid-11,11,11-d3, Undecanoic-11,11,11-D3 acid
InChI formel InChI=1 S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCC(=O)O
IUPAC navn 11,11,11-trideuterioundecanoic acid
Molekylvægt (g/mol) 189.31
Formel vægt 189.181 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity