Learn More
BINDER™ Uncontrolled Dry-Air Purge
For Use With: MKT 115
Brand:  BINDER™ 8012-1805
Additional Details : Weight : 8.00000kg
Product Code. 15904292
  • 44339.78 DKK / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Uncontrolled
  • Includes connection
  • Suitable for preventing condensation
Specifications
Specifications
Dry-air purge - uncontrolled, incl. connection; for preventing condensation
BINDER Chambers MKT 115
Dry-air Purge
Connection