missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ UNC 8732
Beskrivelse
UNC 8732 is a potent nuclear receptor binding SET domain-containing 2 (NSD2) Degrader (DC50 = 60 nM; Dmax = 97%). NSD2 degradation is driven by the aldehyde metabolite of UNC 8732 in vitro. Recruits FBX022 and SCF complexes for degradation to reduce H3K36me2 levels in cells. Reduces viability and restores glucocorticoid sensitivity of NSD2 p.E1099K mutant ALL cells.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[[4-[[[2-(6-Amino-1-oxohexyl)-1,2,3,4-tetrahydro-6-isoquinolinyl]amino]carbonyl]phenyl]methyl]-N-cyclopropyl-3,4-dihydro-3-oxo-2H-1,4-benzoxazine-7-carboxamide trifluoroacetate |
| CAS | 2929304-06-1 |
| Mængde | 5 mg |
| Mål | Active Degraders |
| Molekylær formel | C35H39N5O5.CF3CO2H |
| Renhed | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?