Få mere at vide
Umifenovir, TRC
Artikelnummer. 30455132
Skift visning
Mængde:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455132 5 mg
30451492 100 mg
30450608 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455132 Leverandør Toronto Research Chemicals Leverandørnr. TRCU8501855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 131707-25-0
Molekylær formel C22H25BrN2O3S
Molekylvægt (g/mol) 477.41
Synonym 6-Bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-[(phenylthio)methyl]-1H-indole-3-carboxylic Acid Ethyl Ester
IUPAC navn ethyl 6-bromo-4-[(dimethylamino)methyl]-5-hydroxy-1-methyl-2-(phenylsulfanylmethyl)indole-3-carboxylate
SMIL CCOC(=O)c1c(CSc2ccccc2)n(C)c3cc(Br)c(O)c(CN(C)C)c13
Tekniske data
Kemisk navn eller materiale Umifenovir
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C22H25BrN2O3S/c1-5-28-22(27)20-18(13-29-14-9-7-6-8-10-14)25(4)17-11-16(23)21(26)15(19(17)20)12-24(2)3/h6-11,26H,5,12-13H2,1-4H3
Formel vægt 476.0769
Procent renhed >95%