missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UCL 1684
GREENER_CHOICE

Product Code. 18095592
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18095592 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18095592 Supplier Tocris Bioscience™ Supplier No. 1310/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent blocker of KCa2

Highly potent, non-peptidic blocker of the apamin-sensitive Ca2+-activated K+ channel (KCa2.1) (IC50 = 3 nM in rat sympathetic neurons). Blocks hKCa2.1 and rKCa2.2 channels expressed in HEK 293 cells with IC50 values of 762 and 364 pM respectively.
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale UCL 1684
CAS 199934-16-2
Mængde 5 mg
Molekylær formel C34H30Br2N4
Formel vægt 654.44
Renhed >97%
Molekylvægt (g/mol) 654.45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.