Learn More
UCB9608, MedChemExpress
MedChemExpress UCB9608 is a potent, selective and orally active PI4KIIIβ inhibitor, with an IC50 of 11 nM, selective over PI3KC2 α, β, and γ lipid kinases. UCB9608 improves metabolic stability and exhibits excellent pharmacokinetic profile, acts as a potent immunosuppressive agent.

POA - 3021.37 DKK
Specifications
Chemical Name or Material | UCB9608 |
---|---|
CAS | 1616413-96-7 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C20H26N8O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490947
![]() |
Medchem Express
HY-112613 |
50 mg |
POA
50mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16484658
![]() |
Medchem Express
HY-112613 |
10 mg |
3021.37 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16457377
![]() |
Medchem Express
HY-112613 |
5 mg |
1973.96 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16455496
![]() |
Medchem Express
HY-112613 |
100 mg |
POA
100mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
UCB9608 is a potent, selective and orally active PI4KIIIβ inhibitor, with an IC50 of 11 nM, selective over PI3KC2 α, β, and γ lipid kinases. UCB9608 improves metabolic stability and exhibits excellent pharmacokinetic profile, acts as a potent immunosuppressive agent.Specifications
UCB9608 | |
Off-White | |
C20H26N8O2 | |
NC1=NC(N2CCN(C(NC3=CC=C(OC)C=C3C)=O)C[C@@H]2C)=C4C(N(C)N=C4)=N1 | |
410.47 | |
Research | |
COVID-19-immunoregulation |
1616413-96-7 | |
Research | |
DMSO : 65 mg/mL (158.36 mM; Need ultrasonic) | |
410.47 | |
99.43% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.