missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ UBE2V2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15913605
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15913605 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15913605 Leverandør Invitrogen™ Leverandørnr. PA577091

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The differentiation and activation of monocytes (MO) and monocytic cells is modulated by 1alpha,25-dihydroxyvitamin D3 (Vitamin D3). Recently, using mRNA Differential Display technology, Fritsche J, et al have identified a gene which is induced in freshly isolated human blood MO that are cultured for 4 hours with Vitamin D3. This cDNA encodes a putative 16.5 kD protein. The product of this gene has structural homology with yeast MMS2p and human CROC-1), which defines a structurally and functionally conserved Ubc-like protein family. This protein was called Ubiquitin E2 variant (UBE2V2) or MMS2 (methyl methanesulfonate sensitive 2). The yeast UEV protein, Mms2, is required together with Ubc13 for RAD6/RAD18-dependent postreplicative DNA repair in yeast. These proteins also play a central role in the assembly of K63-linked polyubiquitin chains (4,5) UEV/Ubc complexes have been implicated in the assembly of Lys63-linked polyubiquitin chains that act as a novel signal in postreplicative DNA repair and IkBa kinase activation. Recent crystal structure analysis provides direct evidence that the Mms2/Ubc13 heterodimer is necessary for DNA repair.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen UBE2V2
Applikationer Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2V2
Genadgangsnr. Q15819, Q7M767, Q9D2M8
Gene Alias 1 alpha,25-dihydroxyvitamin D3-inducible; 1110021H13Rik; 4632410D19Rik; 5730524P06Rik; 6820402M05; AI848315; C81524; DDVit 1; DDVIT1; DDVit-1; EDAF-1; EDPF1; EDPF-1; enterocyte differentiation promoting factor; enterocyte differentiation-associated factor 1; enterocyte differentiation-associated factor EDAF-1; enterocyte differentiation-promoting factor 1; meth; methyl methanesulfonate sensitive 2; methyl methanesulfonate sensitive 2, S. cerevisiae, homolog of; Mms2; MMS2 homolog; ubc-like protein MMS2; UBE2V2; ubiquitin conjugating enzyme E2 V2; ubiquitin conjugating enzyme E2 variant 2; ubiquitin-conjugating enzyme E2 variant 2; ubiquitin-conjugating enzyme E2v2; ubiquitin-conjugating enzyme variant MMS2; UEV2; UEV-2; vitamin D3-inducible protein
Gensymboler UBE2V2
Værtsarter Rabbit
Immunogen Recombinant full length Human UBE2V2.
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 287927, 70620, 7336
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.