missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
UBE2T Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38343AF532
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
UBE2T Polyclonal antibody specifically detects UBE2T in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
| UBE2T | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29089 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| Alexa Fluor 532 | |
| Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human UBE2T (NP_054895.1).,, Sequence:, MQRASRLKRELHMLATEPPPGITCWQDKDQMDDLRAQILGGANTPYEKGVFKLEVIIPERYPFEPPQIRFLTPIYHPNIDSAGRICLDVLKLPPKGAWRPSLNIATVLTSIQLLMSEPNPDDPLMADISSEFKYNKPAFLKNARQWTEKHARQKQKADEEEMLDNLPEAGDSRVHNSTQKRKASQLVGIEKKFHPDV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion