Learn More
UBE2D4/Ubch5d Antibody, Allophycocyanin, R&D Systems™
Product Code. 18611916 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18611916 0.1 mL 0.1mL
1 options
Product Code. 18611916 Supplier Novus Biologicals Supplier No. NBP300095APC
 Restricted Item
Forventet forsendelse: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
UBE2D4/Ubch5d Polyclonal specifically detects UBE2D4/Ubch5d in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mL
Research Discipline Cell Biology
Product Type Antibody
Product Line Novus