Learn More
uvex U-Cap Sport Bump Cap - Long Brim
Product Code. 17259221 Shop All Uvex Products
Change view
Click to view available options
Color:
Black
Size:
52 to 54 cm
55 to 59 cm
60 to 63 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Unit Size
17259221 Black 60 to 63 cm Each
17239221 Black 52 to 54 cm Each
17249221 Black 55 to 59 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17259221 Supplier uvex Supplier No. 9794402
  • ONLINE EXCLUSIVE PRICE
    205.00 DKK
    188.12 DKK / Pakke med 1
    Spare 16.88 DKK
    8% Slukket
Forventet forsendelse: 04-12-2026
An innovative bump cap with a hard shell element and honeycomb structure to absorb shock, in addition to greatly reducing the risk of head injuries. Long brim design provides increased sun protection.
  • Ideal for areas where helmets are not compulsory, but there is still a risk of head injuries
  • Innovative hard shell element in armadillo-design, honeycomb structure, and shock-absorbing elements
  • Reliable impact protection in accordance with EN 812
  • Long brim provides increased sun protection
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Product Type Bump Cap
Size 60 to 63 cm
Ventilated Vented
Head Style Baseball Cap