Få mere at vide
DuPont™ Tyvek™ IsoClean™ Sleeves
Shop alle DuPont produkter
:
Size: Universal
Beskrivelse
- White, universal size.
- Tunnelled elastics
- Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
Tekniske data
Tekniske data
| Størrelse | Standard |
| Konstruktion | Flash-spun HDPE |
| Produkttype | Controlled Environment Sleeves |
| Materiale | HDPE |
| Farve | White |
| Sterilitet | Non-sterile |
| Sømtype | Bound |
| Manchet stil | Elastic |
| Model- | IC501B |
| Til brug med (applikation) | Biotech, Pharmaceutical, Medical Device Manufacturing and Electronics Industries, Other Critical or Controlled Environments |
| Vis mere |