Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Small Bag
Designed for the steam sterilization of large or odd shaped pharmaceutical equipment change parts. These bags are closed via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically opened without having to cut or remove the particulate generating autoclave tape.
Brand:  Keystone Cleanroom Products BGT2A0410D
Additional Details : Weight : 0.45720kg
 View more versions of this product
Product Code. 17787887
  • 24720.00 DKK / Pack of 500
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclavable
887 mL
Drawstring
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
10 x 4 in.
25.4 cm
10 cm
30 oz.
COC
White
Steam Sterilization
Tyvek
Steam
<125°C
10 in.
4 in.