Learn More
DuPont™ Tyvek™ 600 Hooded Coverall, Plus Size
Hooded coveralls that are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
8470.00 DKK - 9430.00 DKK
Specifications
Color White
Closure Type Zipper
Seam Type Stitched, Taped
Certifications/Compliance (Europe) (EU) 2016/425, EN 14126, EN 1149-5
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17484943
View Documents
DuPont™
D14981422
4X-Large
8470.00 DKK
Pack of 100
Only null left
Estimated Shipment: 28-07-2025
Add to basket
17494943
View Documents
DuPont™
D14981437
5X-Large
8825.00 DKK
Pack of 100
Only null left
Estimated Shipment: 28-07-2025
Add to basket
17404953
View Documents
DuPont™
D14981445
6X-Large
9120.00 DKK
Pack of 100
Only null left
Estimated Shipment: 28-07-2025
Add to basket
17414953
View Documents
DuPont™
D14981458
7X-Large
9430.00 DKK
Pack of 100
Description
Description
  • Thumb loops
  • Stitched and over-taped seams for protection and strength
  • Tyvek™ zipper and zipper flap for enhanced protection
  • Face, wrist and ankle tunnelled elastication, for good fit and contamination/ linting reduction
Specifications
Specifications
White
Zipper
Stitched, Taped
(EU) 2016/425, EN 14126, EN 1149-5
Documents
Product Certifications
Product Content Correction
Product Title
DuPont™ Tyvek™ 600 Hooded Coverall, Plus Size