Learn More
DuPont™ Tyvek™ 500 TY198S ES
Hooded coverall with ergonomic-protective design. Stitched external seams. Elasticated wrists, ankles and face.
Brand:  DuPont™ D15570297
 View more versions of this product
Product Code. 30305954
  • 2108.30 DKK / Pack of 25
This item is not returnable. View return policy
Description
Description
  • DuPont™ Tyvek™ 500 ES. Hooded coverall available in white, in sizes SM to 3X. Robust yet lightweight. 3-piece hood for optimal fit to head and face when turning. Elasticated face, wrists and ankles as well as glued-in waist elastic. Ample crotch area for freedom of movement. Large, easy-to-grasp zipper puller.
  • Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek™ is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated on the inside. Silicone non-added.
  • Applications of Tyvek™ 500 ES garments include routine tasks in low-risk situations, or when risk is defined, in context of BSL3 or less, and low exposure to body fluids: transport of infected patients, testing, patient triage, refugee camp preparation, people rescue.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hooded Coveralls
Unisex
White
Elastic
Elastic
Elastic
Stitched
Liquid
X-Large
HDPE
Single-use
Elastic
Hooded
Front Zipper with Flap
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek™ 500 TY198S ES > X-Large