Learn More
Tyrphostin B46, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of EGF receptor kinase activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Quantity | 50 mg |
Formula Weight | 322.36 |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Tyrphostin B46 |