missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TYRO3 (RSE), GST Tag Recombinant Protein
Human TYRO3 (RSE), GST Tag Recombinant Protein
4380.00 DKK
Specifications
Accession Number | NP_006284 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 76.6 kDa |
Gene Alias | TYRO3 (RSE), DKT, BRT, TIF |
KinaseFamily | AXL Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11503567
|
Invitrogen
PV3828 |
10μg |
4380.00 DKK
10µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TYRO3 is a receptor tyrosine kinase that has been shown to be involved in signal transduction and cell adhesion.Specifications
NP_006284 | |
76.6 kDa | |
AXL Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
TYRO3 (RSE), DKT, BRT, TIF | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus |