missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TYRO3 (RSE) Recombinant Human Protein
TYRO3 is a receptor tyrosine protein kinase.
13090.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_006284 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 76.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408603
|
Invitrogen™
PR7480A |
100μg |
13090.00 DKK
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10611905
|
Invitrogen™
PV4121 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TYRO3 has been shown to be involved in signal transduction and cell adhesion.
Specifications
NP_006284 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
AXL Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
76.6kDa | |
RUO - research use only | |
TYRO3 (RSE), DKT, BRT, TIF | |
Human | |
GST Tag | |
Frozen |