missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[Tyr0] Neurokinin B, Alfa Aesar™
For cellular and molecular biology applications
Brand: Alfa Aesar J66585.MA
Additional Details : Weight : 0.00010kg
Specifications
1373.61 | |
10mg | |
Tyr-Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2 | |
-30° to -10°C | |
Solid |
HPLC | |
Solid | |
RUO | |
[Tyr0] Neurokinin B |
For Research Use Only