Learn More
Macherey-Nagel™ Type N11 Clear With Label and Scale Snap Cap Vials
Brand:  Macherey-Nagel™ 702713
Additional Details : Weight : 0.01000kg
Product Code. 10094310
  • 123.00 DKK / Pack of 100
Estimated Shipment: 13-05-2024
to see stock.
Specifications
Clear
1.5 mL
Snap Ring, Wide Opening
100 Pack
32 mm
11.6 mm
Label and Scale
100
Flat
100 Pk.