missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 Self Closing
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 34649-061-34628
Additional Details : Weight : 218.16721kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Triple certified: cabinet tested in accordance with EN 14470-1, FM Standard 6050 and UL/ULC Standard 1275
Specifications
1626 mm | |
Safety storage cabinet | |
750 mm | |
Powder-coated textured | |
864 mm | |
225 kg | |
Anthracite Gray, Light Gray | |
620 mm | |
Wing, Left-hinged, Self-closing | |
3 | |
1 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
519 mm | |
Steel | |
620 x 864 x 1947 mm | |
519 x 750 x 1626 mm | |
Standard | |
1947 mm | |
Q30.195.086.FU.WDC | |
90 mm | |
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed |