missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 Self Closing

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 34659-061-34637

Additional Details : Weight : 261.65881kg

 View more versions of this product

Product Code. 15937726

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
  • No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
  • Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Triple certified: cabinet tested in accordance with EN 14470-1, FM Standard 6050 and UL/ULC Standard 1275
Specifications

Specifications

1626 mm
Safety storage cabinet
1050 mm
Powder-coated textured
1164 mm
273.50 kg
Anthracite Gray, Light Gray
620 mm
Wing, Self-closing
3
2
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
519 mm
Steel
620 x 1164 x 1947 mm
519 x 1050 x 1626 mm
Standard
1947 mm
Q30.195.116.FU.WDC
90 mm
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers