missing translation for 'onlineSavingsMsg'
Få mere at vide

Tylosin phosphate, MedChemExpress

Artikelnummer. 16404387
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16404387 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16404387 Leverandør Medchem Express Leverandørnr. HYB0519B

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Tylosin phosphate is a macrolide antibiotic found naturally as a fermentation product of Streptomyces fradiae. Tylosin tartrate exerts potent antimicrobial activity against Gram-positive bacteria. Tylosin phosphate is widely used as a feed additive for promoting animal growth. Tylosin phosphate is used for veterinary purposes against bacterial dysentery and respiratory diseases in poultry, pigs and cattle.

Tylosin phosphate is a macrolide antibiotic found naturally as a fermentation product of Streptomyces fradiae. Tylosin tartrate exerts potent antimicrobial activity against Gram-positive bacteria. Tylosin phosphate is widely used as a feed additive for promoting animal growth. Tylosin phosphate is used for veterinary purposes against bacterial dysentery and respiratory diseases in poultry, pigs and cattle.

Tekniske data

Kemisk navn eller materiale Tylosin phosphate
Fysisk form Powder
CAS 1405-53-4
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C46H80NO21P
Til brug med (applikation) Neuroscience-Neuromodulation
Opløselighedsinformation DMSO : 100 mg/mL (98.61 mM; Need ultrasonic)
SMIL C[C@@H](O[C@@]1([H])O[C@H]([C@H]([C@@H](CC2=O)O)C)[C@H](C[C@H](C(/C=C/C(C)=C/[C@H](CO[C@H](O[C@H](C)[C@@H](O)[C@H]3OC)[C@@H]3OC)[C@@H](CC)O2)=O)C)CC=O)[C@H]([C@@H]([C@H]1O)N(C)C)O[C@@](O[C@@H](C)[C@@H]4O)([H])C[C@]4(O)C.O=P(O)(O)O
Molekylvægt (g/mol) 1014.1
Formel vægt 1014.1
Procent renhed 98.08%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.