Få mere at vide
Tylosin 3-Acetate, TRC
Artikelnummer. 17942125
Skift visning
Mængde:
0.5 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17942125 0.5 mg
17943585 5 mg
17954035 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17942125 Leverandør Toronto Research Chemicals Leverandørnr. TRCT9476600.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63409-10-9
Molekylær formel C48H79NO18
Molekylvægt (g/mol) 958.14
Synonym Tylosin 3-acetate, Tylosin, 3-acetate, Oxacyclohexadecane, tylosin deriv., 3-O-Acetyltylosin, [4R-(4R*,5S*,6S*,7R*,9R*,11E,13E,15R*,16R*)]-4-Acetoxy-15-[[(6-deoxy-2,3-di-O-methyl-β-D-allopyranosyl)oxy]methyl]-6-[[3,6-dideoxy-4-O-(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-3-(dimethylamino)-β-D-glucopyranosyl]oxy]-16-ethyl-5,9,13-trimethyl-2,10-dioxooxacyclohexadeca-11,13-diene-7-acetaldehyde
IUPAC navn [(4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxymethyl]-5,9,13-trimethyl-2,10-dioxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-4-yl] acetate
SMIL CC[C@H]1OC(=O)C[C@@H](OC(=O)C)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(=C\[C@@H]1CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC)\C
Tekniske data
Kemisk navn eller materiale Tylosin 3-Acetate
Mængde 0.5 mg
InChI formel InChI=1S/C48H79NO18/c1-14-34-32(23-60-47-44(59-13)43(58-12)39(54)27(5)62-47)19-24(2)15-16-33(52)25(3)20-31(17-18-50)41(26(4)35(64-30(8)51)21-36(53)65-34)67-46-40(55)38(49(10)11)42(28(6)63-46)66-37-22-48(9,57)45(56)29(7)61-37/h15-16,18-19,25-29,31-32,34-35,37-47,54-57H,14,17,20-23H2,1-13H3/b16-15+,24-19+/t25-,26+,27-,28-,29+,31+,32-,34-,35-,37+,38-,39-,40-,41-,42-,43-,44-,45+,46+,47-,48-/m1/s1
Formel vægt 957.5297