Learn More
Saint-Gobain Tygon Food & Beverage Tubing
Tygon Food and Beverage Tubing is ideal for use in clean-in-place and steam-in-place cleaning and sterilization systems. It is compatible with virtually all commercial cleaners and sanitizers and can be repeatedly autoclaved for up to five cycle times without affecting its overall service life.
Brand:  Saint-Gobain R6F12-23
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12356929
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Wide chemical resistance to both acids and bases thermoplastic elastomer good gas and water vapour impermeability beige -40°C to 135°C temperature range 9.6mm x 1.6mm Norprene
Specifications
Specifications
Autoclavable
Tygon
15 m
Flexible
135°C
9.5 mm
12.7 mm
-60°C
0.48 bar