Learn More
Masterflex™ Tygon™ F-4040-A Fuel and Lubricant Tubing
Resists the swelling and hardening caused by hydro-carbon based fluids. Tygon™ F-4040-A Fuel and Lubricant Tubing routinely handles gasoline, kerosene, heating oils, cutting compounds and glycol-based coolants.
575.00 DKK - 4815.00 DKK
Specifications
Product Type | Industrial Tubing |
---|---|
Material | Tygon F4040 |
For Use With (Application) | Transfer of oil, oil products, diesel, kerosene, hot oil, cutting oil, glycol-based coolant. |
Min. Operating Temperature (English) | -40°F |
Min. Operating Temperature (Metric) | -40°C |
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Thickness (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Thickness (Metric) | Price | Quantity & Availability | |||
11707399
|
Masterflex™
95633-00 |
2.4 mm | 4.8 mm | 1.2 mm |
660.00 DKK
15m |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
11967875
|
Masterflex™
95606-48 |
2.79 mm | 4.76 mm | 0.85 mm |
575.00 DKK
Pack of 12 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
12323870
|
Masterflex™
070533-09L |
1.02 mm | 2.72 mm | 0.85 mm |
4815.00 DKK
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
Description
Features and Benefits
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
Typical Applications
- Small engine fuel lines
- General automotive
- Recreational vehicles
- Lawn and garden equipment
- Coolant transfer
- Heating fuels
- Cutting compounds
- Polishing equipment
- Lubrication lines
Specifications
Industrial Tubing | |
Transfer of oil, oil products, diesel, kerosene, hot oil, cutting oil, glycol-based coolant. | |
-40°C | |
158°F |
Tygon F4040 | |
-40°F | |
Yellow | |
70°C |