missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Tygon™ Extension Tube

Suitable for standard applications. Heidolph™ Tygon™ Extension Tube is durable and resistant.

Brand:  Heidolph™ 525-60024-00

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10458781

  • 69.32 DKK / Each


Estimated Shipment
02-06-2021
Add to basket

Description

Description

  • Non-toxic.
  • Non-oxidizing.
  • Good resistance to acids, bases and inorganic media.
  • Very low gas permeability.
  • Good performance life.
Specifications

Specifications

FDA (21 CFR 177.2601) and GLP
75°C
Good
0.89 mm
2.69 mm
Extension Tubing
Non-toxic, non-oxidizing, good resistance to acids, bases and inorganic media, very low gas permeability, good performance life.
0.89mm I.D./2.69mm O.D.
7.0 m Water
Segregation of plasticizers is possible.
Transparent
Continuous/short time 0.5/1.5 bar
Autoclavable
Tygon
0.9 mm
Pump
-50°C
Autoclave for 30min at 1bar and 120┬░C (material may change color) or with ethylene oxide.
Very low
Videos
SDS
Documents