Få mere at vide
DuPont™ Tychem™ NT430
Shop alle DuPont produkter
Størrelse:
2X-Large, 11
Large, 9
Medium, 8
Small, 7
X-Large, 10
X-Small, 6
Pakningsstørrelse:
12 par
Beskrivelse
- 0.23 mm nitrile gloves for jobs requiring chemical performance plus optimum fit, feel and comfort
- Lightweight,'Second-Skin' feel and low-modulus formulation reduces hand fatigue
- CE CAT III - EN ISO 374-1:2016/TYPE A, JKOPT
- EN ISO 374-5:2016
- Food contact compatible
Tekniske data
Tekniske data
| Certificeringer/overholdelse (Europa) | CE CAT III |
| Farve | Blue |
| Materiale | Nitrile |
| Størrelse | 2X-Large, 11 |
| Tykkelse (engelsk) | 9 mil |
| Tykkelse (metrisk) | 0.23 mm |
| Model- | D15536298 |
- Store away from light and humidity
- Rinse gloves in running water before removing, using a neutral detergent if necessary
- Gloves should not be used around heat, open flame, sparks or in potentially flammable environments
- For Professional Use Only