Få mere at vide
Tybamate, LoGiCal
Artikelnummer. 30460514
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460514 10 mg
1 options
Artikelnummer. 30460514 Leverandør LoGiCal Leverandørnr. LGCFOR1626.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tybamate
CAS 4268-36-4
Analyt- eller komponentnavne Tybamate
Forsendelsestilstand Room Temperature
Molekylær formel C13 H26 N2 O4
Certificeringer/compliance ISO 9001
Synonym Tybamate, N-Butylcarbamic acid 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester, Butylcarbamic acid 2-(hydroxymethyl)-2-methylpentyl ester carbamate (ester), Butylcarbamic acid 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester, Butylcarbamic acid ester with 2-(hydroxymethyl)-2-methylpentyl carbamate, 2-Methyl-2-propyltrimethylene butylcarbamate carbamate, Benvil, Depran, Effisax, N-Butyl-2-methyl-2-propyl-1,3-propanediol dicarbamate, N-n-Butyl-2-methyl-2-propyl-1,3-propanediol dicarbamate, NSC 172126, Nospan, Reposan, Solacen, Solacin, Tibamate, Tibamax, Tybatran, W 713
InChI formel InChI=1S/C13H26N2O4/c1-4-6-8-15-12(17)19-10-13(3,7-5-2)9-18-11(14)16/h4-10H2,1-3H3,(H2,14,16)(H,15,17)
Løsningstype Neat
SMIL CCCCNC(=O)OCC(C)(CCC)COC(=O)N
IUPAC navn [2-(carbamoyloxymethyl)-2-methylpentyl] N-butylcarbamate
Industry Type Clinical Testing
Molekylvægt (g/mol) 274.36
Mængde 10 mg
Formel vægt 274.1893 g/mol