Learn More
Lenz Laborglasintrumente Two-Neck Round-Bottom Flasks, Side Neck Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 03204170
This item is not returnable.
View return policy
Description
- Distillation flasks
- With parallel side neck
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifications
1 L | |
Duran Glass | |
Side Neck NS 14/23 | |
131 mm |
Product Content Correction
Product Title
Lenz Laborglasintrumente Two-Neck Round-Bottom Flasks, Side Neck Parallel > 1000 mL
Spot an opportunity for improvement?Share a Content Correction