Learn More
Eppendorf™ 96-Well Twin.tec™ LoBind™ PCR Plates, Skirted
Skirted 96-Well Twin.tec™ LoBind™ PCR Plates allow you to recover nearly 100% of your DNA/RNA molecules, and with no surface coating, there is no risk of sample contamination. Batch-tested and certified free from DNA, DNase, RNase and PCR inhibitors, they are perfect for PCR.
Brand:  Eppendorf™ 0030129539
Additional Details : Weight : 0.76000kg
Product Code. 15851525
  • 1555.00 DKK / Pack of 25
Estimated Shipment: 21-05-2024
to see stock.
Description
Description
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack™ matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Specifications
Specifications
PCR Clean
8 x 12
Clear
Polypropylene, Polycarbonate
Non-sterile
Yes
25
PCR Clean
Colorless
Yes
Autoclavable
Yes
96
Real Time PCR
2250 x g