Learn More
Eppendorf™ 96-Well Twin.tec™ LoBind™ PCR Plates, Skirted
Skirted 96-Well Twin.tec™ LoBind™ PCR Plates allow you to recover nearly 100% of your DNA/RNA molecules, and with no surface coating, there is no risk of sample contamination. Batch-tested and certified free from DNA, DNase, RNase and PCR inhibitors, they are perfect for PCR.
Brand: Eppendorf™ 0030129539
Additional Details : Weight : 0.76000kg
Description
- Maximum recovery rates for improved assay results
- Available with barcode (upon request)
- Ideal for quantitative real-time PCR as well
- Exceptionally solid and torque-resistant polycarbonate frame
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Specifications
PCR Clean | |
8 x 12 | |
Clear | |
Polypropylene, Polycarbonate | |
Non-sterile | |
Yes | |
25 | |
PCR Clean | |
Colorless |
Yes | |
Autoclavable | |
Yes | |
96 | |
Real Time PCR | |
2250 x g | |
Yes | |
Yes |