Få mere at vide
TUSC4 Antibody [Janelia Fluor« 646], Novus Biologicals Biologicals™
Artikelnummer. 30498734 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde Pakningsstørrelse
30498734 0.1 mL 0.1ml
1 options
Artikelnummer. 30498734 Leverandør Novus Biologicals Leverandørnr. NBP335395JF646
 Begrænset vare
Forventet forsendelse: 12-10-2026
Rabbit Polyclonal Antibody
TUSC4 Polyclonal antibody specifically detects TUSC4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen TUSC4
Applikationer ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Klassifikation Polyclonal
Konjugeret Janelia Fluor 646
Formulering 50mM Sodium Borate
Gene Alias G21 protein, Gene 21 protein, homologous to yeast nitrogen permease (candidate tumor suppressor), nitrogen permease regulator 2-like protein, nitrogen permease regulator-like 2 (S. cerevisiae), NPR2, NPR2L2810446G01Rik, NPRL2, Tumor suppressor candidate 4NPR2-like protein, TUSC4
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human TUSC4 (NP_006536.3).,, Sequence:, MGSGCRIECIFFSEFHPTLGPKITYQVPEDFISRELFDTVQVYIITKPELQNKLITVTAMEKKLIGCPVCIEHKKYSRNALLFNLGFVCDAQAKTCALEPIVKKLAGYLTTLELESSFVSMEESKQKLVPIMTILLEELNASGRCTLPIDESNTIHLKVIEQRPDPPVAQEYDVPVFTKDKEDFFNSQWDLTTQQILPYIDGFRHIQKISAEADVELNLV
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Cell Cycle and Replication
Primær eller sekundær Primary
Gen-id (Entrez) 10641
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG