Learn More
RETSCH Tungsten Carbide O-ring
Brand:  RETSCH 05.114.0068
16824.50 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.01000kg
Product Code. 10155143
  • 17710.00 DKK / Each
Estimated Shipment: 28-08-2024
to see stock.
Specifications
O-Ring
1Each
Tungsten Carbide
For Vibratory Disc Mill RS 200 50mL Grinding Set