Få mere at vide
Tulobuterol Hydrochloride, TRC
Artikelnummer. 30452104
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452104 5 g
30452995 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30452104 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8972555G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56776-01-3
Molekylær formel C12 H18 Cl N O . Cl H
Molekylvægt (g/mol) 264.19
Synonym 2-Chloro-α-[[(1,1-dimethylethyl)amino]methyl]-benzenemethanol Hydrochloride (1:1), Benzenemethanol, 2-chloro-α-[[(1,1-dimethylethyl)amino]methyl]-, hydrochloride (9CI), Atenos, Berachin, Brelomax, Bremax, C 78, Chlibamol, Hokunalin, Lobuterol, Respacal, Tulobuterol hydrochloride, Ventaire
IUPAC navn 2-(tert-butylamino)-1-(2-chlorophenyl)ethanol;hydrochloride
SMIL Cl.CC(C)(C)NCC(O)c1ccccc1Cl
Tekniske data
Kemisk navn eller materiale Tulobuterol Hydrochloride
Mængde 5 g
InChI formel InChI=1S/C12H18ClNO.ClH/c1-12(2,3)14-8-11(15)9-6-4-5-7-10(9)13;/h4-7,11,14-15H,8H2,1-3H3;1H
Formel vægt 263.0844