Få mere at vide
Tulobuterol-d9 Hydrochloride, TRC
Artikelnummer. 30454647
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30454647 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454647 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8972522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tulobuterol tert-butyl-d9 hydrochloride
CAS 1325559-14-5
Renhedsgrad noter HPLC
Molekylær formel C12 2H9 H9 Cl N O . Cl H
Mængde 2.5 mg
Synonym 1-(2-Chlorophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol hydrochloride, Tulobuterol D9 (tert-butyl D9) hydrochloride
InChI formel InChI=1S/C12H18ClNO.ClH/c1-12(2,3)14-8-11(15)9-6-4-5-7-10(9)13;/h4-7,11,14-15H,8H2,1-3H3;1H/i1D3,2D3,3D3;
SMIL Cl.[2H]C([2H])([2H])C(NCC(O)c1ccccc1Cl)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1-(2-chlorophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol;hydrochloride
Molekylvægt (g/mol) 273.25
Formel vægt 272.1409
Procent renhed >95