missing translation for 'onlineSavingsMsg'
Learn More

Tugsten solution for ICP 10g/L HNO3/HF matrix, Fisher chemical™

Catalog Number p-7084018
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10±0.02g/L
Packaging Bottle
Quantity 100 mL
Type Mass Spectrometry Standard
Analyte or Component Names W
Solvent or Matrix HNO3/HF
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Tungsten (W) 10000mg/l in HNO3 2%,HF 5%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'