Learn More
Fisherbrand™ Tube Rack for Shaking Incubators
Fisherbrand™ Tube Rack is designed to be used with a shaking incubator and can hold a variety of tube sizes.
Brand:  Fisherbrand™
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 16393226
  • 1270.00 DKK / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • For use with shaking incubator
  • Constructed of plastic, polypropylene, aluminum, or wire
  • Designed to hold tubes
Specifications
Specifications
Vertical
15 mL, 50 mL
Plastic
32, 20
15 mL, 50 mL Tubes
Tube Rack
16313226
1 Ea.