Learn More
Fisherbrand™ Tube Rack for Shaking Incubators
Fisherbrand™ Tube Rack is designed to be used with a shaking incubator and can hold a variety of tube sizes.
Brand:  Fisherbrand™
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 16303246
  • 1365.00 DKK / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • For use with shaking incubator
  • Constructed of plastic, polypropylene, aluminum, or wire
  • Designed to hold tubes
Specifications
Specifications
Vertical
50 mL
Polypropylene
15
50 mL Tubes
Tube Rack
16313226
1 Ea.