missing translation for 'onlineSavingsMsg'
Få mere at vide

BOLA Tube Fittings T

Artikelnummer. 10489761
Skift visning
Klik for at se tilgængelige muligheder
Diameter (metrisk) Udvendig:
6,35 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Diameter (metrisk) Udvendig
10489761 6,35 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10489761 Leverandør BOLA Leverandørnr. D50506
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

Tube Fittings T PTFE, nut made of glass-fibre reinforced PTFE

  • Tube fitting T-shaped made of PTFE
  • Three connections with nuts made of glass-fibre reinforced PTFE.
  • Universal chemical resistance
  • The product is only exposed to PTFE.

Tekniske data

Certificeringer/compliance FDA
Autoklaverbar Autoclavable
Diameter (metrisk) Udvendig 6,35 mm
Højde (metrisk) 39 mm
Farve White
Fleksibilitet Rigid
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.